Polygodial ≥98% (by HPLC)

Supplier: Enzo Life Sciences

BML-CA242-0005 BML-CA242-0025
89156-824EA 192.76 CAD
89156-824 89156-826
Polygodial ≥98% (by HPLC)
Polygodial

TRPA1 activator.


  • Formulation: White to off white solid or waxy solid.


Produced from the bark of Drymis winteri. Naturally occurring drimane sesquiterpene found in water pepper (Polygonum hydropiper) which has been used in folk medicine for a variety of indications. Potent and selective activator of TRPA1 (EC₅₀ = 400 nM).


Soluble in DMSO (25 mg/mL) or ethanol (25 mg/mL).


Synonym: (1R,4aS,8aS)-1,4,4a,5,6,7,8,8a-Octahydro-5,5,8a-trimethyl-1,2-naphthalenedicarboxaldehyde

Formula: C₁₅H₂₂O₂
Storage Temperature: Freezer
MDL Number: MFCD00238664
CAS Number: 6754-20-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6bb5925c07d3c0680d89748803045570f71b7d06&vid=988f7a2c7976128106223dae1a3fab55f6d5942d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Source Isolated from Polygonum hydropiper
Purity ≥98% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR