HC-030031 ≥98% (by HPLC)
Supplier: Enzo Life Sciences
BML-CA241-0010
BML-CA241-0050
89156-820EA
176.96
CAD
89156-820
89156-822
HC-030031 ≥98% (by HPLC)
HC-030031
A potent and selective TRPA1 inhibitor.
- TRPA1 inhibitor
White solid.
Soluble in DMSO (25 mg/ml).
Formula:
C₁₈H₂₁N₅O₃ Storage Temperature: Ambient |
CAS Number:
349085-38-7 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...