BCTC ≥98%

Supplier: Enzo Life Sciences

BML-CA231-0010 BML-CA231-0050
89156-796EA 178.54 CAD
89156-796 89156-798
BCTC ≥98%
BCTC

TRPV1 antagonist


BCTC is a potent and selective inhibitor of capsaicin-induced (IC50=35 nM) and acid-induced (IC50=6 nM) VR1 activation. It displays analgesic activity in rat models of inflammatory and neuropathic pain. TRPV1 antagonist.

Formula: C₂₀H₂₅ClN₄O
MDL Number: MFCD08690556
CAS Number: 393514-24-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3d0e7d83e81e098a2590b65aa29c6e904048ca2b&vid=607afe9c5a7f18e6886edafe8e747bc50da4e7bf&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR