SKF-96365 hydrochloride ≥98% (by HPLC)

Supplier: Enzo Life Sciences

Synonyms: 1-(2-(4-Methoxyphenyl)-2-(3-(4-methoxyphenyl)propoxy)ethyl)-1H-imidazole hydrochloride, 1-[2-(4-Methoxyphenyl)-2-[3-(4-methoxyphenyl)propoxy]ethyl]-1H-imidazole hydrochloride, SKF-96365

BML-CA230-0005 BML-CA230-0025
89156-792EA 237 CAD
89156-792 89156-794
SKF-96365 hydrochloride ≥98% (by HPLC)
SKF-96365 hydrochloride

A novel, selective inhibitor of receptor-mediated calcium entry. It inhibits receptor-mediated calcium entry in stimulated platelets (IC50=8-12µM) neutrophils and endothelial cells at concentrations which do not affect internal Ca2+ release. Note, in some cells SK&F 96365 can activate a novel Ca2+ entry pathway. Blocks TRPC3 and TRPC6 activation. Inhibits the histamine-induced formation of nitric oxide in human endothelial cells.


  • TRP channel blocker


White solid.


Soluble in water (30mg/ml).

Formula: C₂₂H₂₆N₂O₃·HCl
MW: 402.9 g/mol
Melting Pt: 117…119 °C
Storage Temperature: Ambient
MDL Number: MFCD00236407
CAS Number: 130495-35-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=595ec74985f2c8cb04d535a62dc17c97fc466415&vid=4e0bc3539b1abef976424aee19775235835de2e6&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR