Niguldipine hydrochloride ≥98%
Supplier: Enzo Life Sciences
BML-CA216-0010 BML-CA216-0050
89156-782EA 172.22 CAD
89156-782 89156-784
Niguldipine hydrochloride ≥98%
Niguldipine hydrochloride
Ca2+ channel blocker
A classic L-type Ca2+ channel blocker (IC50=75nM) which can also inhibit T-type Ca2+ channels (IC50=244nM). It is a novel inhibitor of drug transport by p-glycoprotein (both stereoisomers are equipotent at inhibition of p-glycoprotein).
CAS Number: 119934-51-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0eca35128aba9cfff9bb1e4aede619580ec7f550&vid=b8ffa3cbb3b47338ae3f9bccd3d41fd30cbbeee9&prid=web&clid=SAPDAM