Anti-CK8 Mouse Monoclonal Antibody [clone: 2A4]

Supplier: Enzo Life Sciences
CA1141-0100
89156-766EA 427 CAD
89156-766
Anti-CK8 Mouse Monoclonal Antibody [clone: 2A4]
Antibodies
Host: Mouse, Isotype: IgG1

  • For Immunocytochemistry, Western Blot

Recognizes human, mouse, rat, pig cytokeratin proteins 8, 18 and 19 respectively. Detects bands of 52 kDa, 45 kDa and 40 kDa in Western Blot.

Type: Primary
Antigen: CK8
Clonality: Monoclonal
Clone: 2A4
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR