Anti-CGRP Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
CA1134-0025 CA1134-0100
89156-750EA 249.64 CAD
89156-750 89156-752
Anti-CGRP Rabbit Polyclonal Antibody
Antibodies
Host: Rabbit

  • For Immunohistochemistry

Recognizes human, mouse and rat CGRP.

Type: Primary
Antigen: CGRP
Clonality: Polyclonal
Clone: 0
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR