Guanfacine hydrochloride ≥99% (by TLC)

Supplier: Enzo Life Sciences
Danger

BML-AR102-0100 BML-AR102-0500
89156-616EA 142.2 CAD
89156-616 89156-618
Guanfacine hydrochloride ≥99% (by TLC)
Guanfacine hydrochloride

Adrenergic receptor ligand.


  • Formulation: White solid.


Centrally acting α2-adrenergic receptor agonist. In the guinea pig cerebral cortex, the Kd is 20 and 880 nM for the α2A- and the α2C-adrenoceptor, respectively. Antihypertensive agent.


Soluble in water (25 mg/mL).

Formula: C₉H₉Cl₂N₃O
MDL Number: MFCD00798230
CAS Number: 29110-48-3
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=97dde2ff8cecdc88a0570ec3872c446e64b28d3c&vid=16c050548ac91d4940d3b4d1efc4e22b03176481&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity ≥99% (HPLC)
Identity Determined by NMR


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR