Bopindolol malonate ≥97% (by HPLC)
Supplier: Enzo Life Sciences
|
|
Synonyms:
Wandonorm, SCHEMBL348872, HE393689, CTK8F831, LS-120081, D01794, AC1LCS45
BML-AR100-0100
BML-AR100-0500
89156-608EA
139.04
CAD
89156-608
89156-610
Bopindolol malonate ≥97% (by HPLC)
Bopindolol malonate
Adrenergic receptor ligand.
- Formulation: White solid.
A slowly dissociating β1-adrenergic receptor antagonist. Antihypertensive. Antioxidant (IC₅₀ = 1.8 µM) and free radical scavenging activity.
Soluble in water at 3 mg/mL.
Formula:
C₂₃H₂₈N₂O₃·C₃H₄O₄ MW: 484.54 g/mol |
MDL Number:
MFCD00798228 CAS Number: 62658-64-4 |
Specification Test Results
Purity | ≥97% (HPLC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...