Bleomycin sulfate (mixture) ≥98% (by HPLC)
Supplier: Enzo Life Sciences
BML-AP302-0010
BML-AP302-0050
89156-580EA
526.14
CAD
89156-580
89156-582
Bleomycin sulfate (mixture) ≥98% (by HPLC)
Bleomycin sulfate (mixture)
Anti-tumor agent
- White crystalline solid
Induces apoptosis via DNA damage. Radiomimetic DNA-cleaving agent that produces double and single DNA strand breaks through an oxygen-radical-dependent mechanism. Inhibits intracellular DNA replication. DNA alkylating agent.
Soluble in water (20 mg/mL) or methanol
Packaging: Shipping temperature: Ambient
Formula:
C₅₅H₈₄N₁₇O₂₁S₃·xH₂SO₄ MW: 1512.6 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00070310 CAS Number: 9041-93-4 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...