Sulindac sulfone ≥94% (by HPLC)

Supplier: Enzo Life Sciences

Synonyms: Z-5-Fluoro-2-methyl-1-[p-(methyl-sulphonyl)benzylidene]indene-3-acetic acid, Z-5-Fluoro-2-methyl-1-[p-(methyl-sulfonyl)benzylidene]indene-3-acetic acid

BML-AP201-0005 BML-AP201-0025
89156-576EA 142.2 CAD
89156-576 89156-578
Sulindac sulfone ≥94% (by HPLC)
Sulindac sulfone

Induces apoptosis but does not inhibit cyclooxygenase. Inhibits growth and induces apoptosis in human cancer cell lines. Metabolite of Sulindac.


  • Sulindac metabolite


Yellow solid.


Soluble in DMSO (>25mg/ml) or 100% ethanol (3mg/ml, warm).

Formula: C₂₀H₁₇FO₄S
MW: 372.4 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00871143
CAS Number: 59864-04-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b4ef08c8a6a635ba4df05615da70bf040fa150b6&vid=8f801bc9374f5caedb750ec94cc26bfda1809d25&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR