Sulindac Sulfide ≥99% (by TLC)

Supplier: Enzo Life Sciences

BML-AP200-0005 BML-AP200-0025
89156-572EA 139.04 CAD
89156-572 89156-574
Sulindac Sulfide ≥99% (by TLC)
Sulindac Sulfide

Active metabolite of sulindac. Inhibits cyclooxygenase. More selective for PGHS-1 (ID50=0.5 µM) over PGHS-2 (ID50=14 µM). Induces apoptosis by a cyclooxygenase-independent mechanism. Inhibits colorectal cancer growth and γ-secretase. NF-κB inhibitor. Inhibits Ras signaling.


  • COX inhibitor


Yellow-orange solid.


Soluble in DMSO (>25mg/ml) or 100% ethanol (5mg/ml, warm).

Formula: C₂₀H₁₇FO₂S
Storage Temperature: Ambient
MDL Number: MFCD00869764
CAS Number: 32004-67-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bd5f4b4b712d28cdb3293b123c24d115aedac39f&vid=367edeeedc8ce589e0aa7a5f5df1c050a4ceb93b&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR