4-Amino-1,8-naphthalimide ≥96% (by NMR)
Supplier: Enzo Life Sciences
BML-AP101-0020
BML-AP101-0100
89156-564EA
178.54
CAD
89156-564
89156-566
4-Amino-1,8-naphthalimide ≥96% (by NMR)
4-Amino-1,8-naphthalimide
PARP inhibitor
Potent inhibitor of poly (ADP-ribose) polymerase-1 (PARP-1) (IC50=0.18μM). Reduces ischemia-reperfusion injury in the heart and skeletal muscle.
Formula:
C₁₂H₈N₂O₂ |
MDL Number:
MFCD00006921 CAS Number: 1742-95-6 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...