Alamethicin ≥98% (by TLC)

Supplier: Enzo Life Sciences

BML-A150-0005 BML-A150-0025
89156-320EA 364.98 CAD
89156-320 89156-322
Alamethicin ≥98% (by TLC)
Alamethicin

Mixture of alamethicin homologs


  • White to off-white solid


Membrane permeabilizing antibiotic. Applications include increasing [32P] incorporation into phosphatidylinositol 4-phosphate and permeabilization of sarcoplasmic reticulum vesicles. Causes fusion of lipid vesicles.


Soluble in DMSO, 100% ethanol or methanol


Packaging: Shipping temperature: Ambient

Formula: C₉₂H₁₅₀N₂₂O₂₅
Storage Temperature: Refrigerator
MDL Number: MFCD00151517
CAS Number: 59588-86-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1a93b448d7a09e2798c7a2c95cdc5ddba67bc17d&vid=799753961ebd3c157e29a97614bbbe0f79f22dcc&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR