SCREEN-WELL® REDOX Library, Enzo Life Sciences
Supplier: Enzo Life Sciences
BML-2835-0100 BML-2835-0500
89156-302EA 2005.02 CAD
89156-302 89156-304
SCREEN-WELL® REDOX Library, Enzo Life Sciences
Microplates Immunology Microplates and Strips
The Screen-Well™ REDOX library contains 84 compounds with defined prooxidant or antioxidant activity.
Compounds are dissolved in DMSO at 10mM and aliquoted into deep-well plates at 100 or 500µL per well
Stable for one year from the date of receipt when stored at -80°C and shipped on Dry Ice.
A variety of structurally and mechanistically different compound classes are included such as hydroperoxides, polyphenolics, metal chelators, thiols, thiol traps, radical scavengers, lazaroids and glutathione modulators as well as small molecule enzyme mimetics for SOD and glutathione peroxidase. The library is a useful tool for studying the roles of pro- and antioxidant molecules in cells as well as for use in in vitro applications.