SCREEN-WELL® Protease Inhibitor Library, Enzo Life Sciences
Supplier: Enzo Life Sciences
The Screen-Well™ Protease Inhibitor Library includes 53 known protease inhibitors on a single 96-well plate. Inhibitors are supplied dissolved in DMSO at 10 mM and aliquoted into deep-well plates at either 100 or 500 microliters per well.
The library includes inhibitors to a broad range of important proteases and is a convenient and cost-effective way to purchase a panel of protease inhibitors for chemical genomics, assay development, and other applications. It includes inhibitors of: Calpains, TPPII, Neutrophil Elastase, Cathepsins, DPPIV, Caspases, Granzyme B, gamma-Secretase, Furin, MMPs, ACE, Kallikrein, Proteasome, Aminopeptidase B, and many more.