SCREEN-WELL® Nuclear Receptor Ligand Library, Enzo Life Sciences
Supplier: Enzo Life Sciences
BML-2802-0100 BML-2802-0500
89156-256EA 3344.86 CAD
89156-256 89156-258
SCREEN-WELL® Nuclear Receptor Ligand Library, Enzo Life Sciences
Microplates Immunology Microplates and Strips
The SCREEN-WELL® Nuclear Receptor Ligand Library contains 74 compounds with defined, putative and potential activity at nuclear receptors.
Receptor agonists and antagonists are included
Stable for one year from the date of receipt when stored at -80°C and shipped on Dry Ice.
The library consists of a chemically diverse group of compounds which are supplied dissolved in DMSO at 10 mM (1mM for 5 compounds) and aliquoted into deep-well, 96-well plates at 500 µL or 100 µL per well. The library is an ideal tool for chemical genomics, receptor de-orphaning and other routineMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable