Cell Counting Kit-F, Enzo Life Sciences
Supplier: Enzo Life Sciences
850-245-KI01
850-245-KI02
89156-010EA
553
CAD
89156-010
89156-012
Cell Counting Kit-F, Enzo Life Sciences
Assays
Cellular Assays
Fluorometric kit for simple and accurate cell proliferation and cytotoxicity assays
Advantages includes no radioisotopes, more sensitive than the colorimetric assay, short staining reaction time, no solubilization step, correlates with the [3H]-thymidine incorporation assay.