APO-BRDU™ kit, TUNEL Assay
Supplier: Enzo Life Sciences
850-042-KI01
89155-906EA 1280.01 CAD
89155-906
APO-BRDU™ kit, TUNEL Assay
Assays Cellular Assays
Kit for measuring apoptosis with high sensitivity by Dual Color Flow Cytometry in mammalian cells. Br-dUTP is more readily incorporated into the genome of apoptotic cells than are larger ligands like fluorescein, biotin, etc. This greater incorporation coupled with the use of a fluorescein labelled anti-BrdU monoclonal antibody (Prod. No. ALX-804-197L) gives rise to a stronger signal in flow cytometry.
Ordering information: Quantity: 50 assays. Contents: Contains all the reagents necessary, including positive and negative control cells.