Cell Counting Kit-8, Enzo Life Sciences
Supplier: Enzo Life Sciences
850-039-0100 850-039-KI02 850-039-KI01
89155-900EA 1050 CAD
89155-900 76199-140 89155-898
Cell Counting Kit-8, Enzo Life Sciences
Assays Cellular Assays
Colorimetric kit for simple and accurate cell proliferation and cytotoxicity assays.
Advantages: One-step, ready-to-use solution, no radioisotopes, no solubilization step, high sensitivity, correlates with the [3H]-thymidine incorporation assay. Can be added directly to the cell media for fast, high-throughput screening without a solubilization process obtaining highly reproducible and accurate results.