DNA (Deoxyribonucleic acid) (from calf thymus) ≥99%, purified

Supplier: Enzo Life Sciences

840-040-C010
89155-846EA 154 CAD
89155-846
DNA (Deoxyribonucleic acid) (from calf thymus) ≥99%, purified
DNA (Deoxyribonucleic acid)

Purified from calf thymus and activated by digestion with DNAse I.


For use with PARP-1 or PARP-2 in vitro activity tests


Concentration: 0,2 mg/ml in TRIS-EDTA.

Storage Temperature: Refrigerator
MDL Number: MFCD00130922
CAS Number: 9007-49-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR