Anti-CD40 Rat Monoclonal Antibody

Supplier: Enzo Life Sciences
805-046-C100 805-046-C500
89155-292EA 395 CAD
89155-292 89155-294
Anti-CD40 Rat Monoclonal Antibody
Antibodies
Host: Rat, Isotype: IgG2a

Recognizes mouse CD40.

Type: Primary
Antigen: CD40
Clonality: Monoclonal
Clone: 0
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR