Anti-MRP4 Rat Monoclonal Antibody

Supplier: Enzo Life Sciences
801-038-C075 801-038-C150
89153-256EA 925.88 CAD
89153-256 89153-258
Anti-MRP4 Rat Monoclonal Antibody
Antibodies
Host: Rat, Isotype: IgG2a

  • For Immunocytochemistry, Immunohistochemistry, Western Blot

Recognizes human and mouse MRP4.

Type: Primary
Antigen: MRP4
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR