Anti-MRP2 Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
801-037-C125 801-037-C250
89153-252EA 976.44 CAD
89153-252 89153-254
Anti-MRP2 Mouse Monoclonal Antibody
Antibodies
Host: Mouse, Isotype: IgG2b

  • For Immunocytochemistry, Immunohistochemistry, Western Blot

Recognizes an internal epitope of human, mouse and rat MRP2. Does not cross-react with human P-glycoprotein, MDR3 P-glycoprotein, MRP1, MRP3 or MRP5. Does not recognize mouse MRP2 in immunohistochemistry.

Type: Primary
Antigen: MRP2
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR