Anti-MRP1 Rat Monoclonal Antibody

Supplier: Enzo Life Sciences
801-007-C125 801-007-C250
89153-172EA 946.42 CAD
89153-172 89153-174
Anti-MRP1 Rat Monoclonal Antibody
Antibodies
Host: Rat, Isotype: IgG2a

  • For Immunocytochemistry, Immunohistochemistry, Western Blot

Recognizes the internal epitope (G238SDLWSLNKE247) of human and mouse MRP1, overexpressed in various human non-P-glycoprotein MDR tumor cell lines. Does not cross-react with human P-glycoprotein, MDR3 P-glycoprotein or human MRP2 (cMOAT) to MRP 5.

Type: Primary
Antigen: MRP1
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR