Polyinosinic-polycytidylic acid potassium salt (synthetic), TLRgrade™

Supplier: Enzo Life Sciences

746-021-M002 746-021-M005
89153-078EA 254.38 CAD
89153-078 89153-080
Polyinosinic-polycytidylic acid potassium salt (synthetic), TLRgrade™
Polyinosinic-polycytidylic acid potassium salt

Activator for TLR3 and MDA5.


Specific activator for TLR3 (Toll-like receptor 3) and MDA5/Helicard.


Contains polymer, residual salt and water. Sufficient for at least 200 cellular activation assays at 10 – 25 µg/mL.

Formula: (C₁₀H₁₀N₄NaO₇P)x·(C₉H₁₁N₃NaO₇P)x
Storage Temperature: Refrigerator
CAS Number: 31852-29-6

Order Now

Specification Test Results

Source Synthetic
Purity  Activity and endotoxin tested - TLRgrade™
Endotoxin Content <0.002 EU/µg
Formulation Lyophilized, sterile


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR