HT-2 Toxin ≥98% (by TLC)

Supplier: Enzo Life Sciences
Danger

630-113-M001 630-113-M005
89153-024EA 474 CAD
89153-024 89153-026
HT-2 Toxin ≥98% (by TLC)
HT-2 Toxin

Mycotoxin.


  • Formulation: White to slight yellow solid.


Trichothecene group mycotoxin. Induces apoptosis.


Soluble in 100% ethanol, DMSO or ethyl acetate. Slightly soluble in petroleum ether, poorly soluble in water.


Synonym: 15-Acetoxy-3α,4β-dihydroxy-8α-(3-methylbutyryloxy)-12,13-epoxytrichothec-9-ene.

Formula: C₂₂H₃₂O₈
Storage Temperature: Freezer
CAS Number: 26934-87-2
UN: 2811
ADR: 6.1,I
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=07d42ca3944f684a3267d80eb6d1bf7fa75f1e8d&vid=b27088a8265a9c8acac6c688fc5372429c7c0509&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Source Semisynthetic, derived from T2 toxin from Fusarium tricinctum
Purity ≥98% (TLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR