Ochratoxin A ≥98% (by HPLC, TLC)
Supplier: Enzo Life Sciences
|
Danger
|
630-089-M001
630-089-M005
630-089-M025
89152-972EA
105.05
CAD
89152-972
89152-974
89152-976
Ochratoxin A ≥98% (by HPLC, TLC)
Ochratoxin A
Mycotoxin. Nephrotoxic.
- Formulation: White to off-white crystalline powder.
Mycotoxin. Natural contaminant of mouldy food and feed. It has a number of toxic effects, the most prominent being nephrotoxicity. Immunosuppressive, genotoxic, teratogenic and carcinogenic. Stimulates lipid peroxidation. Induces oxidative DNA lesions coupled with direct DNA adducts via quinone formation.
Isolated from Aspergillus ochraceus.
Soluble in methanol (10 mg/mL).
Formula:
C₂₀H₁₈ClNO₆ Storage Temperature: Refrigerator |
MDL Number:
MFCD00078079 CAS Number: 303-47-9 UN: 2811 ADR: 6.1,II Merck Index: 14,06740 |
|
Specification Test Results
Purity | ≥98% (HPLC, TLC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...