Luminol (3-Aminophthalhydrazide) ≥97% (by HPLC)

Supplier: Enzo Life Sciences
Warning

610-002-G001 610-002-G005
89152-822EA 69.52 CAD
89152-822 89152-824
Luminol (3-Aminophthalhydrazide) ≥97% (by HPLC)
Luminol (3-Aminophthalhydrazide)

Key reagent for generating luminescence


  • Formulation: Off-white to brown solid.


Key reagent for generating luminescence. A reagent for the highly sensitive and selective detection of nitric oxide (NO) in biological samples by a luminol-H2O2 chemiluminescence method.

Formula: C₈H₇N₃O₂
Storage Temperature: Freezer
MDL Number: MFCD00006890
CAS Number: 521-31-3
EINECS: 208-309-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=54efc16eca10e617daa37a8aae830460d978eda9&vid=2fada0f5c254704e45d2abed948a55b066bf1d8e&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR