IPTG (isopropyl-β-D-thiogalactopyranoside) ≥98% (by HPLC)

Supplier: Enzo Life Sciences
Warning

Synonyms: IPTG, (2R,3R,4S,5R,6S)-2-(Hydroxymethyl)-6-(isopropylthio)tetrahydro-2H-pyran-3,4,5-triol, Isopropyl Beta-D-1-thiogalactopyranoside

582-012-G005 582-012-G025
89152-816EA 246.48 CAD
89152-816 89152-818
IPTG (isopropyl-β-D-thiogalactopyranoside) ≥98% (by HPLC)
IPTG (Isopropyl-β-D-thiogalactopyranoside)

β-galactosidase activator.


  • Formulation: White to off-white powder.


Inducer of β-D-galactosidase. Used in the stimulation of β-galactosidase in cellular systems in which dioxane would disrupt normal cell function. Contains no animal-derived products.


Soluble in water (5%) or methanol.

Formula: C₉H₁₈O₅S
MW: 238.31 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00063273
CAS Number: 367-93-1
EINECS: 206-703-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8c5620378fb2fa102f599a111cdfa49a7719c94e&vid=186fdb2ea4135ce1f3d0bd2ba6e387799c429f77&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity ≥98% (HPLC)
Identity Determined by IR

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR