X-Gal (5-bromo-4-chloro-3-indolyl-ß-D-galactopyranoside) ≥98%

Supplier: Enzo Life Sciences

Synonyms: BCIG, X-GAL, (2S,3R,4S,5R,6R)-2-((5-Bromo-4-chloro-1H-indol-3-yl)oxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol

582-002-G001 582-002-G005
89152-806EA 254.38 CAD
89152-806 89152-808 89152-812
X-Gal (5-bromo-4-chloro-3-indolyl-ß-D-galactopyranoside) ≥98%
X-Gal (5-Bromo-4-chloro-3-indolyl-ß-D-galactopyranoside)

Chromogenic substrate for the enzyme β-galactosidase.


  • Formulation: White powder

Formula: C₁₄H₁₅BrClNO₆
MW: 408.63 g/mol
MDL Number: MFCD00005666
CAS Number: 7240-90-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0e24d1b85b17148bb6ce55a2bb25256cabb9191a&vid=eef0bb61dff236661a3cee8522aa9d679685760c&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR