Compound A 98

Supplier: Enzo Life Sciences

Synonyms: 2-((4-acetophenyl)-2-chloro-N-methyl)ethylammonium chloride, 4-(1-Chloro-2-(methylamino)ethyl)phenyl acetate hydrochloride

550-516-M005 550-516-M025
89152-722EA 105.86 CAD
89152-722 89152-724
Compound A 98
Compound A

Glucocorticoid receptor modulator.


  • Formulation: White to off-white solid.


Cell permeable, nonsteroidal glucocorticoid receptor modulator with high binding affinity.


Exerts anti-inflammatory potential by down-modulating TNF-induced pro-inflammatory gene expression, such as IL-6 and E-selectin. Unlike dexamethasone it does not enhance glucocorticoid response element (GRE)-driven gene expression or induce glucocorticoid receptor (GR) binding to GRE-dependent genes in vivo.


Soluble in DMSO, 100% ethanol or water.


Synonym: CpdA; 2-((4-acetophenyl)-2-chloro-N-methyl)ethylammonium chloride

Formula: C₁₁H₁₄ClNO₂·HCl
MW: 264.2 g/mol
Storage Temperature: Ultra-Low Temperature Freezer (–80°C)
MDL Number: MFCD16883076
CAS Number: 14593-25-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ef30a442825ffcdbf0c2141933a502c4d587688c&vid=e46bdd4d9101b93ca88fbdaab635d26dd639cd07&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity (HPLC) ≥98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR