MPEP hydrochloride ≥99%

Supplier: Enzo Life Sciences

Synonyms: 2-Methyl-6-(phenylethynyl)pyridine hydrochloride, MPEP HCl

550-368-M005 550-368-M025
89152-618EA 96.38 CAD
89152-618 89152-620
MPEP hydrochloride ≥99%
MPEP hydrochloride

mGluR5 antagonist.


  • Formulation: Off-white powder.


Potent and selective antagonist for mGluR5 receptor. Systemically active in vivo.


Soluble in water, DMSO or methanol.


Synonym: 6-Methyl-2-(phenylethynyl)pyridine.HCl

Formula: C₁₄H₁₁N.HCl
MW: 229.71 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD02262119
CAS Number: 219911-35-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e6d8bdb66723109e55bcd1397e83f7900f9e61de&vid=901e1949d61faf3228846bc6525cacedc1dce8e1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity ≥99%
Identity Determined by NMR


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR