(Z)-4-Hydroxytamoxifen ≥98%

Supplier: Enzo Life Sciences

Synonyms: (Z)-4-(1-(4-(2-(Dimethylamino)ethoxy)phenyl)-2-phenylbut-1-en-1-yl)phenol, (Z)-4-(1-[4-(Dimethylaminoethoxy)phenyl]-2-phenyl-1-butenyl)phenol, (Z)-4-OHT, trans-4-Hydroxytamoxifen

550-361-M001 550-361-M005
89152-604EA 109.02 CAD
89152-604 89152-606
(Z)-4-Hydroxytamoxifen ≥98%
(Z)-4-Hydroxytamoxifen

Tamoxifen metabolite


Metabolite of tamoxifen (Prod. No. ALX-550-095). Exhibits a more potent estrogen agonist/antagonist activity than the parent drug. Has antioxidant properties. Intramembranous inhibitor of lipid peroxidation.

Formula: C₂₆H₂₉NO₂
MW: 387.51 g/mol
MDL Number: MFCD00278780
CAS Number: 68047-06-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=33077abdc234c4ed09797f676f24d150e849420b&vid=f24c378afe5df620d404693ba381f63030e09518&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR