Serotonin hydrochloride ≥98% (by HPLC)

Supplier: Enzo Life Sciences

Synonyms: 5-Hydroxytryptamine Hydrochloride

550-328-G001 550-328-M050
89152-536EA 395 CAD
89152-536 89152-538 89152-540
Serotonin hydrochloride ≥98% (by HPLC)
Serotonin hydrochloride

Neurotransmitter


  • Formulation: White to gray crystalline powder.


Neurotransmitter. Endogenous 5-HT agonist produced by serotonergic neurons in the CNS and enterochromaffin cells in the gastrointestinal tract.

Formula: C₁₀H₁₃ClN₂O
MW: 212.68 g/mol
MDL Number: MFCD00012686
CAS Number: 153-98-0
UN: 2811
ADR: 6.1 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c3625b5cfb87b2773b03577313448dd49255ab70&vid=5283c202e2137e8604ffbf03c62bb3972ba4aecd&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR