Verapamil hydrochloride ≥99%

Supplier: Enzo Life Sciences

Synonyms: 5-((3,4-Dimethoxyphenethyl)(methyl)amino)-2-(3,4-dimethoxyphenyl)-2-isopropylpentanenitrile hydrochloride, Verapamil HCl, 5-[(3,4-Dimethoxyphenethyl)methylamino]-2-(3,4-dimethoxyphenyl)-2-isopropylvaleronitrile Hydrochloride, (±)-Verapamil hydrochloride

550-306-G001 550-306-G005
89152-492EA 71.1 CAD
89152-492 89152-494
Verapamil hydrochloride ≥99%
Verapamil hydrochloride

Blocks slow voltage-sensitive L-type Ca2+ channels. Adrenergic antagonist. Inhibits platelet-activating factor and PMA-stimulated prostaglandin production in Kupffer cells. Induces apoptosis. In vitro resistance modifier in drug resistant human tumor cell lines.


  • Ca2+ channel inhibitor


White crystalline powder.


Soluble in 100% ethanol (26mg/ml), methanol or water (83mg/ml).

Formula: C₂₇H₃₈N₂O₄
MW: 491.07 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00055208
CAS Number: 152-11-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d6956ee03ed4f049c24f125edb89a1caa9ac00da&vid=35f0d4543259645a49f4b7d1ce2be449a3bb2239&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR