6-Cyano-7-nitroquinoxaline-2,3-dione disodium salt, disodium salt ≥98% (by TLC)

Supplier: Enzo Life Sciences

Synonyms: CNQX disodium salt, CNQX 2Na

550-283-M005 550-283-M010 550-283-M025
89152-464EA 140.62 CAD
89152-464 89152-466 89152-468
6-Cyano-7-nitroquinoxaline-2,3-dione disodium salt, disodium salt ≥98% (by TLC)
6-Cyano-7-nitroquinoxaline-2,3-dione

AMPA/kainate receptor antagonist.


  • Rust red solid.


Water soluble, potent competitive kainate/quisqualate (non-NMDA) receptor antagonist. More soluble form of CNQX (Prod. No. ALX-550-042).


Soluble in water (5 mg/mL).

Formula: C₉H₂N₄O₄.2Na
MW: 276.12 g/mol
Storage Temperature: Freezer
MDL Number: MFCD09953908
CAS Number: 115066-14-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=25953e8d4a173a531fa62b0e45dd2b79cec93190&vid=9fd0baa559c1b1fd5e99ad95ce2e173e2b4a1e0c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity (TLC) ≥98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR