NBQX ≥98% (by HPLC)

Supplier: Enzo Life Sciences
Warning

550-272-M005 550-272-M025
89152-436EA 252.8 CAD
89152-436 89152-438
NBQX ≥98% (by HPLC)
NBQX

Water soluble, potent and selective AMPA/kainate receptor antagonist.


  • Formulation: Red to brown solid.


Soluble in water (5 mg/mL) or DMSO.


Synonyms: 2,3-Dihydroxy-6-nitro-7-sulphamoylbenzo(f)-quinoxaline.2Na, 6-nitro-7-sulphamoylbenzo(f)-quinoxaline-2,3-dione.2Na

Formula: C₁₂H₈N₄O₆S
Storage Temperature: Freezer
MDL Number: MFCD00270054
CAS Number: 118876-58-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=893ad58d00e66cc60cd35f9d1b0a9ba71723ecb6&vid=5ea8cee2bce30f9fd660801b8a962f90e7452c6d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity (HPLC) ≥98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR