Diazoxide ≥98% (by HPLC)

Supplier: Enzo Life Sciences
Warning

550-260-G001 550-260-M500
89152-410EA 483.48 CAD
89152-410 89152-412
Diazoxide ≥98% (by HPLC)
Diazoxide

ATP-sensitive K⁺ channel activator.


  • Formulation: White to off-white powder.


Effective concentrations in patch clamp experiments of 100 to 600 µM. Blocks desensitisation of AMPA receptors. Acts in pancreatic β-cells, where it is more potent than pinacidil. Antihypertensive and vasodilatory agent.


Soluble in DMSO (25 mg/mL); insoluble in water.

Formula: C₈H₇ClN₂O₂S
Storage Temperature: Refrigerator
MDL Number: MFCD00078578
CAS Number: 364-98-7
EINECS: 206-668-1
Merck Index: 14,03004
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8a968e2807921d66fcb1855d3c57f271c269636a&vid=70387b939a0fcd4cac737d5a7b82dd9339a8a4aa&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity (HPLC) ≥98%
Appearance White to off-white powder


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR