Clofilium tosylate ≥97% (by HPLC)
Supplier: Enzo Life Sciences
|
Warning
|
Synonyms:
4-Chloro-N,N-diethyl-N-heptylbenzenebutanaminium . tosylate
550-242-M100
550-242-M250
89152-388EA
1423.58
CAD
89152-388
89152-390
Clofilium tosylate ≥97% (by HPLC)
Clofilium tosylate
K⁺ channel blocker. Antiarrhytmic. Cardiac depressant.
- Formulation: White solid.
Soluble in water, methanol or DMSO.
Formula:
C₂₈H₄₄ClNO₃S Storage Temperature: Ambient |
CAS Number:
92953-10-1 UN: 2811 ADR: 6.1,III |
Specification Test Results
Purity (HPLC) | ≥97% |
Identity | Determined by ¹H-NMR |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...