Bepridil hydrochloride ≥98%
Supplier: Enzo Life Sciences
550-237-M010
550-237-M025
89152-376EA
86.9
CAD
89152-376
89152-378
Bepridil hydrochloride ≥98%
Bepridil hydrochloride
Ca2+ channel inhibitor
Non-selective Ca2+ channel blocker. Stimulates the binding of 1,4-dihydropyridine-based drugs to Ca2+ channels. Inhibits Na+/Ca2+ exchange.
CAS Number:
74764-40-2 |