5-Nitro-6,7-dichloro-1,4-dihydro-2,3-quinoxalinedione ≥98% (by TLC)

Supplier: Enzo Life Sciences

550-228-M005 550-228-M025
89152-362EA 218.04 CAD
89152-362 89152-364
5-Nitro-6,7-dichloro-1,4-dihydro-2,3-quinoxalinedione ≥98% (by TLC)
5-Nitro-6,7-dichloro-1,4-dihydro-2,3-quinoxalinedione

Potent and selective NMDA receptor glycine site antagonist.


  • Formulation: Yellow powder.


Soluble in DMSO or dilute aqueous base.


Synonym: ACEA-1021

Formula: C₈H₃Cl₂N₃O₄
Storage Temperature: Refrigerator
CAS Number: 153504-81-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f6f8defcd49f67bed17175c59d4ef0570ff5406c&vid=600adc11b94186f7d53ae5aac345a77c80dd049e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity (TLC, elemental analysis) ≥98%

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR