Luzindole ≥96%

Supplier: Enzo Life Sciences

Synonyms: N-Acetyl-2-benzyl-tryptamine

550-184-M005 550-184-M025
89152-290EA 143.78 CAD
89152-290 89152-292
Luzindole ≥96%
Luzindole

Competitive melatonin receptor antagonist.


  • Formulation: White to off-white crystalline solid.

Formula: C₁₉H₂₀N₂O
MW: 292.382 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00672498
CAS Number: 117946-91-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0fd5eb30829184b867804f0af5faf408347c139a&vid=8c2e0ff1e0bfbaa7b1e8f786dbc091f0e3e54092&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity ≥96%
Appearance White to off-white crystalline solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR