Capsazepine ≥98%

Supplier: Enzo Life Sciences

Synonyms: N-(4-Chlorophenethyl)-7,8-dihydroxy-4,5-dihydro-1H-benzo[c]azepine-2(3H)-carbothioamide

550-145-M005 550-145-M025
89152-234EA 203.82 CAD
89152-234 89152-236
Capsazepine ≥98%
Capsazepine

Capsaicin antagonist


Analog of capsaicin (Prod. No. BML-EI125-0200) that acts as a capsaicin antagonist. Inhibits resiniferatoxin and capsaicin-induced contractions of guinea pig trachea. Inhibits C-fiber-smooth muscle contractions induced by capsaicin, bradykinin and neurokinin A. Increases intracellular Ca2+ levels.


Synthetic.

Formula: C₁₉H₂₁ClN₂O₂S
MW: 376.90 g/mol
MDL Number: MFCD00153778
CAS Number: 138977-28-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cfd483b0ce98dc80380336b9b30b7a8adf2007a3&vid=ca3ba25ad5479c3aa8c412e7e14901c7c1d36f68&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR