NBQX ≥99% (by HPLC)

Supplier: Enzo Life Sciences
Warning

550-044-M005 550-044-M025
89152-052EA 153.26 CAD
89152-052 89152-054
NBQX ≥99% (by HPLC)
NBQX

Most potent and selective AMPA/kainate receptor antagonist. More soluble form is NBQX disodium salt.


  • Formulation: Yellow crystalline powder.


Soluble in DMSO. A 1 µM aqueous solution may be prepared by taking up in >2.5 equivalents of NaOH and backtitrating to pH 8.0 to 8.5 with concentrated HCl.


Synonyms: 2,3-Dihydroxy-6-nitro-7-sulphamoylbenzo(f)-quinoxaline; 6-nitro-7-sulphamoylbenzo(f)-quinoxaline-2,3-dione

Formula: C₁₂H₈N₄O₆S
Storage Temperature: Freezer
MDL Number: MFCD00270054
CAS Number: 118876-58-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=07b9c832cde53ac5c0a8c3d7728ec40a7b60a9dd&vid=680ddbf510cb6a71e19d1549fb524d6085d029c1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity (HPLC) ≥99%

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR