LY-83583 ≥95%

Supplier: Enzo Life Sciences

550-002-M005 550-002-M025
89151-978EA 178.54 CAD
89151-978 89151-980
LY-83583 ≥95%
LY-83583

Guanylyl cyclase inhibitor


Competitive inhibitor of soluble guanylyl cyclase (sGC) (IC50=2µM) and of cGMP production. Neurotoxin. Blocks antigen-induced leukotriene release and reduces platelet-endothelial cell adhesion. Blocks acethylcholine-induced vasorelaxation.


Synthetic.

Formula: C₁₅H₁₀N₂O₂
CAS Number: 91300-60-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=69c9729a2d6ec00e1c3b8799dcb29d38ad0203a6&vid=e8d65feac14b3a237a2f1384fda15f985987f764&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR