Human Recombinant IL33 (from E. coli)

Supplier: Enzo Life Sciences

522-098-3010 522-098-C010
89151-862EA 998.56 CAD
89151-862 89151-864
Human Recombinant IL33 (from E. coli)
Proteins and Peptides

Produced in E. coli. Untagged human IL-33 (aa 112-270).

Order Now

SPECIFICATIONS

Protein/peptide name IL33 IL33
Environmentally Preferable
Protein/peptide type Recombinant Recombinant
Species Human Human
Source E. coli E. coli
Endotoxin content <0.1EU/µg purified protein (LAL test; Bio Whittaker).
Biological activity Activates human ST2-dependent NF-κB pathway. Does not activate mouse ST2-dependent NF-κB.
Purity >95% (SDS-PAGE)
Concentration 0.1mg/ml after reconstitution.
Formulation Lyophilized. Contains PBS.
Reconstitution instructions Reconstitute with 100µl sterile distilled water. Further dilutions should be made with medium containing 5% fetal calf serum.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR