Human Recombinant IL-1beta (soluble) (from E. coli)

Supplier: Enzo Life Sciences

522-056-3010 522-056-C010
89151-792EA 1249.78 CAD
89151-792 89151-794
Human Recombinant IL-1beta (soluble) (from E. coli)
Proteins and Peptides

Produced in E. coli. Human IL-1β (interleukin-1β) (aa 117-270) is fused to a linker peptide (10 aa) and an N-terminal FLAG®-tag.

Order Now

SPECIFICATIONS

Protein/peptide name IL-1beta (soluble) IL-1beta (soluble)
Environmentally Preferable
Protein/peptide type Recombinant Recombinant
Species Human Human
Source E. coli E. coli
Endotoxin content <0.1EU/µg purified protein (LAL test; Bio Whittaker).
Biological activity Activates the NF-κB pathway.
Purity >90% (SDS-PAGE)
Concentration 0.1mg/ml after reconstitution.
Formulation Lyophilized. Contains PBS.
Reconstitution instructions Reconstitute with 100µl sterile water.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR