Human Recombinant CD134L (soluble)
Supplier: Enzo Life Sciences
522-023-3010 522-023-C010
89151-724EA 1098.1 CAD
89151-724 89151-726
Human Recombinant CD134L (soluble)
Proteins and Peptides
Produced in HEK 293 cells. The extracellular domain of human CD134L (OX40L) (aa 52-183) is fused at the N-terminus to a linker peptide (8 aa) and a FLAG®-tag.