Human Recombinant TRAIL (soluble) (from E. coli)
Supplier: Enzo Life Sciences
522-003-3010 522-003-C010
89151-678EA 998.56 CAD
89151-678 89151-680
Human Recombinant TRAIL (soluble) (from E. coli)
Proteins and Peptides
Produced in E. coli. The extracellular domain of human TRAIL (aa 95-281) is fused at the N-terminus to a linker peptide (8 aa) and a FLAG®-tag.